Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Cytosine 500 G
SDP

Supplier:  Research Products International Corp C55000500.0

Encompass_Preferred

Molecular Formula: C4H5N3O
Molecular Weight: 111.1
CAS Number: 71-30-7

Catalog No. 50-223-0289


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.