Learn More
Medchemexpress LLC Cytosine | 71-30-7 | 99.99% | 111.10 | 5 G
SDP
Supplier:  Medchemexpress LLC HYI06265G
Encompass_Preferred
Cytosine is one of the four main bases found in DNA and RNA. Its modifications exhibit circadian oscillations involved in epigenetic diversity and aging. It can be used in cancer research.
  • Endogenous metabolite
  • Cytosine modifications exhibit circadian oscillations that are involved in epigenetic diversity and aging
  • Can be used in cancer research
Catalog No. 50-004-30767