Learn More
Medchemexpress LLC Cytosine | 71-30-7 | 99.99% | 111.10 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYI0626500MG
Encompass_Preferred
Cytosine is one of the four main bases found in DNA and RNA. Cytosine modifications exhibit circadian oscillations that are involved in epigenetic diversity and aging. Cytosine can be used in cancer research.
  • Endogenous metabolite
  • Modifications exhibit circadian oscillations involved in epigenetic diversity and aging
  • Can be used in cancer research
Catalog No. 50-004-28556