Learn More
AdipoGen Cytostatin (250 µg)
SDP
Supplier:  AdipoGen AGCN20093C250
Encompass_Preferred
Chemical. CAS: 457070-06-3 | 682329-63-1 (free base). Formula: C21H32NaO7P. MW: 450.4. Isolated from Streptomyces sp. MJ654-NF4. Potent and selective protein phosphatase 2A (PP2A) inhibitor. Cell adhesion inhibitor. Inhibits the adhesion of B16 melanoma cells to laminin and collagen type IV in a dose dependent manner but not to fibronectin. Antitumor agent. Antimetastatic. Apoptosis inducer.
Catalog No. 50-205-3453