Learn More
Medchemexpress LLC HY-111138 10mg Medchemexpress, CZC-8004 CAS:916603-07-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11113810MG
Encompass_Preferred
Medchemexpress, HY-111138 10mg CZC-8004 CAS:916603-07-1 CZC-8004 is a pan-kinase inhibitor and binds a range of tyrosine kinases, including ABL kinase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8826