Learn More
Selleck Chemical LLC D- E2962-25MG
SDP
Supplier:  Selleck Chemical LLC E296225MG
Encompass_Preferred
D-( )-Malic acid (D-Malic acid) found in apples and many other fruits and plants is an active enantiomer of Malic acid It is a competitive inhibitor of L(-)malic acid transport
Catalog No. 50-000-40132