Learn More
Medchemexpress LLC D-4-Hydroxyphenylglycine | 22818-40-2 | 100.0% | 167.16 | 500 G

Supplier: Medchemexpress LLC HY34588500G
D-4-Hydroxyphenylglycine is a crucial raw material used in the production of semisynthetic β-lactam antibiotics. It is intended for research use only.
- Crucial raw material for semisynthetic β-lactam antibiotics.
- White to off-white solid appearance.
- Soluble in DMSO, requiring ultrasonic, warming, and heating to 80°C.
- Powder storage: -20°C for 3 years, 4°C for 2 years.
- In solvent storage: -80°C for 6 months, -20°C for 1 month.
- Functions as an antibiotic.
- Involved in the anti-infection pathway.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.