Learn More
Medchemexpress LLC D-4-Hydroxyphenylglycine | 22818-40-2 | MFCD00004262 | 99.99% | 167.16 | 100 G
SDP
Supplier:  Medchemexpress LLC HY34588100G
Encompass_Preferred
D-4-Hydroxyphenylglycine is a crucial raw material employed in the synthesis of semisynthetic β-lactam antibiotics, including Amoxicillin and Cefadroxil. It is intended for research use only.
  • Used in the production of semisynthetic β-lactam antibiotics.
  • For research use only.
Catalog No. 50-004-03728