Learn More
Cayman Chemical D-Ala-D-Ala 1g
SDP
Supplier:  Cayman Chemical 396481 G
Encompass_Preferred
A dipeptide moiety of bacterial peptidoglycan; its replacement with D-Ala-D-lactate in peptidoglycan reduces the affinity of vancomycin and is responsible for vancomycin resistance
Catalog No. 50-291-8338