Learn More
Medchemexpress LLC HY-111956B 10mg Medchemexpress, D-Ala-Lys-AMCA (hydrochloride) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY111956B10MG
Encompass_Preferred
Medchemexpress, HY-111956B 10mg D-Ala-Lys-AMCA (HCl) CAS: D-Ala-Lys-AMCA hydrochloride is a known proton-coupled oligopeptide transporter 1 (PEPT1) substrate that emits blue fluorescence. D-Ala-Lys-AMCA hydrochloride may be transported into liver cancer cells and Caco-2 cells based on fluorescence analysis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8991