Learn More
Medchemexpress LLC [D-Ala2, D-Leu5]-enkephalin | 63631-40-3 | 99.9% | 569.65 | C29H39N5O7 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10534325MG
Encompass_Preferred
BW-180C (DADLE) is a synthetic enkephalin-family peptide that functions as a selective delta-opioid receptor (DOR) agonist with reported neuroprotective activity. Provided as a research-grade solid for preclinical and in vitro studies, it reversibly inhibits neuronal transcription without causing cell injury.
  • Selective delta-opioid receptor agonist peptide.
  • Exhibits neuroprotective activity and reversible inhibition of neuronal transcription.
  • High purity suitable for research applications.
  • White to off-white solid form for easy handling and storage.
  • Molecular weight 569.65 and formula C29H39N5O7 for identification.
  • Available in small-scale quantities suitable for laboratory use.
  • Stable under low-temperature storage when handled according to guidance.
Catalog No. 50-002-43919