Learn More
Medchemexpress LLC [D-Ala2, D-Leu5]-enkephalin | 63631-40-3 | MFCD00076411 | 99.9% | 569.65 g/mol | C29H39N5O7 | 5 MG

Supplier: Medchemexpress LLC HY1053435MG
BW-180C (DADLE; [D-Ala2, D-Leu5]-enkephalin) is a synthetic pentapeptide δ-opioid receptor agonist used in preclinical research to study opioid signaling, neuroprotection, and tissue ischemia-reperfusion models. Supplied as a high-purity solid, it is intended for laboratory research use only and should be handled under appropriate safety and storage conditions.
- High purity suitable for research applications (≈99.9%).
- Molecular formula C29H39N5O7 and molecular weight 569.65 g/mol.
- Solid powder form for convenient dosing and storage.
- Used in cellular and tissue models of neuroprotection and ischemia-reperfusion injury.
- Target: delta-opioid (DOR) receptor activity.
- Recommended sealed storage at low temperatures to preserve stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.