Learn More
Medchemexpress LLC [D-Ala2]-GIP (human) | 444073-04-5 | 99.0% | 4983.60 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP32471MG
Encompass_Preferred
[D-Ala2]-GIP (human) is a GIP receptor agonist (EC50 of 630 pM) that improves glucose tolerance and shows neuroprotective activity in Parkinson's disease models. It also enhances cognitive function and hippocampal synaptic plasticity in obese diabetic rats, making it suitable for research on type 2 diabetes and Parkinson's disease.
  • GIP receptor agonist with an EC50 of 630 pM
  • Improves glucose tolerance by stimulating insulin release
  • Demonstrates neuroprotective activity in MPTP-induced Parkinson's disease models
  • Enhances cognitive function and hippocampal synaptic plasticity in obese diabetic rats
  • Suitable for research on type 2 diabetes and Parkinson's disease
Catalog No. 50-003-98689