Learn More
Medchemexpress LLC D-Alanine | 338-69-2 | MFCD00008077 | 99.92% | 89.09 | 100 G

Supplier: Medchemexpress LLC HY41700100G
D-Alanine is a weak GlyR (inhibitory glycine receptor) and PMBA agonist, with an EC50 of 9 mM for GlyR. It induces dose-dependent depolarization of motoneurones in newborn rats. In vivo, it relieves Dizocilpine-induced prepulse inhibition deficits in mice when combined with CBIO.
- Weak GlyR (inhibitory glycine receptor) and PMBA agonist.
- Induces dose-dependent depolarization of motoneurones in newborn rats.
- Relieves Dizocilpine-induced prepulse inhibition deficits in mice in vivo.
- Purity: 99.92%.
- Appearance: Solid, white to off-white.
- Shipping: Room temperature in continental US; may vary elsewhere.
- Storage: Powder -20°C for 3 years, 4°C for 2 years; In solvent -80°C for 2 years, -20°C for 1 year.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.