Learn More
Medchemexpress LLC D-Alanine, 4,4,5,5-tetradehydro-D-norvalyl-, 2,2,2-trifluoroacetate (1:1) | 87156-01-2 | 95.0% | 298.22 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY147097A
Encompass_Preferred
EDA-DA TFA is a N-terminally tagged dipeptide probe that can be utilized to label Peptidoglycan (PG) of bacteria. It functions as a click chemistry reagent, possessing an Alkyne group that enables it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. This product is intended for research use only.
  • N-terminally tagged dipeptide probe
  • Labels peptidoglycan (PG) of bacteria
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules
Catalog No. 50-003-64001