Learn More
Medchemexpress LLC D-alanine ethyl ester hydrochloride | 6331-09-5 | MFCD00190725 | 98.0% | 153.61 g/mol | C5H12ClNO2 | 50 G

Supplier: Medchemexpress LLC HYW01735050G
D-alanine ethyl ester hydrochloride is the hydrochloride salt of the ethyl ester of D-alanine. It is used as an amino acid derivative and synthetic building block in research, including peptide synthesis and studies of stereochemistry and enzymatic reactions. The compound is supplied as a solid for laboratory use and should be stored protected from moisture for stability.
- Molecular formula: C5H12ClNO2
- Molecular weight: 153.61 g/mol
- White powder solid, reported melting point ~78-83 °C
- Storage: keep sealed at 4 °C; in solution store at -20 °C to -80 °C for extended stability
- Applications: amino acid derivative, peptide synthesis building block, stereochemical studies
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.