Learn More
Medchemexpress LLC D-alanine, N-[(9H-fluoren-9-ylmethoxy)carbonyl]-N-methyl | 138774-92-2 | MFCD00153385 | 99.9% | 325.36 g/mol | C19H19NO4 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW022281100G
Encompass_Preferred
Fmoc-N-Me-D-Ala-OH is an Fmoc-protected N-methyl D-alanine derivative used as a building block in peptide synthesis and related research. Supplied as a solid powder with high chemical purity, it is intended for incorporation into synthetic peptides and for method development in medicinal chemistry and peptide chemistry workflows.
  • High purity (99.9%) suitable for research-grade synthesis.
  • Fmoc-protected N-methyl D-alanine for incorporation into peptides.
  • Solid powder form for easy weighing and handling.
  • Stable when stored under recommended conditions for reliable use.
  • Compatible with standard solid-phase peptide synthesis protocols.
  • Available in gram-scale quantities for laboratory-scale synthesis.
Catalog No. 50-003-00446