Learn More
Medchemexpress LLC D-alanyl-D-alanine | 923-16-0 | MFCD00066038 | 97.0% | 160.17 g/mol | C6H12N2O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW015450R10MG
Encompass_Preferred
D-Ala-D-Ala (D-alanyl-D-alanine) is an analytical reference standard used in research and method development. It represents the terminal dipeptide of bacterial peptidoglycan and is used as a calibration and reference compound in chromatography and mass spectrometry workflows.
  • Used as an analytical reference standard for chromatography and mass spectrometry.
  • Represents the terminal dipeptide in bacterial peptidoglycan, relevant to microbiology studies.
  • High purity suitable for analytical workflows (≥97.0%).
  • Known molecular weight 160.17 g/mol and chemical formula C6H12N2O3.
  • Available in small pack sizes suitable for laboratory use.
Catalog No. 50-003-02789