Learn More
Medchemexpress LLC D-Alanyl-D-alanine | 923-16-0 | MFCD00066038 | 98.0% | 160.17 g/mol | C6H12N2O3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW015450100MG
Encompass_Preferred
D-Ala-D-Ala is the D-alanyl-D-alanine dipeptide found at the terminal position of bacterial peptidoglycan peptide chains. It is used as a biochemical reagent for studies of cell wall biosynthesis, peptidoglycan structure, and interactions with antibiotics.
  • Occurs as the peptidoglycan terminal dipeptide in bacteria
  • Enables studies of cell wall biosynthesis and transpeptidation
  • Useful in assays probing antibiotic binding and resistance mechanisms
  • Solid, white to off-white powder with defined storage conditions
  • High purity (98.0%) suitable for biochemical research
  • Molecular weight 160.17 g/mol and formula C6H12N2O3
Catalog No. 50-003-02550