Learn More
Bioassay Systems EnzyChrom™ D-Amino Acid Assay Kit. Quantitative determination of D-Amino Acids in biological and other samples.
SDP
Supplier:  Bioassay Systems EDAA100
Encompass_Preferred
Quantitative determination of D-Amino Acids in biological and other samples. Key Features: Fast and sensitive. Linear detection range: 0.86 to 500 µM (colorimetric assay) and 0.18 to 50 µM (fluorimetric assay) for 60 min reaction. Convenient. The procedure involves adding a single working reagent and reading after 60 minutes. High-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated to process thousands of samples per day. Method: OD570nm or FL530/585nm; Samples: Tissue, milk, and other biological samples; Size: 100 tests.
Catalog No. NC1829705