Learn More
Medchemexpress LLC D-arabino-hex-1-enitol, 1,5-anhydro-2-deoxy-3,4,6-tris-O-(phenylmethyl) | 55628-54-1 | MFCD00061640 | 99.8% | 416.51 g·mol^-1 | C27H28O4 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW061616
Encompass_Preferred
Tri-O-benzyl-D-glucal is a benzyl-protected glucal used as a glycosylation building block and reagent in carbohydrate synthesis and glycobiology research. It is supplied as a solid and is typically used as a protected glycal donor in glycosylation reactions and oligosaccharide assembly.
  • High purity: 99.82% (HPLC) reported on batch Certificate of Analysis.
  • Useful as a glycosylation donor for carbohydrate synthesis.
  • Solid form, convenient for weighing and handling in synthetic workflows.
  • Molecular formula C27H28O4 and molecular weight 416.51 g·mol^-1.
  • Available in small-scale quantities (for example, 250 MG) for research use.
Catalog No. 50-002-08474