Learn More
Medchemexpress LLC D-Arabinopyranose | 28697-53-2 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYN7082500MG
Encompass_Preferred
D-Arabinopyranose is an analytical standard intended for research and analytical applications as a laboratory chemical. This monosaccharide shows strong growth inhibition against *Caenorhabditis elegans* with an IC50 of 7.5 mM.
  • Used as a reference standard in qualitative, quantitative, and methodological research
  • Suitable for experiments in HPLC, GC, and MS
  • Appears as a white to off-white solid
  • Initial source is microorganisms
  • For research use only
  • Melting/freezing point: 162-164 °C
  • Relative density: 1.1897 g/cm³
Catalog No. 50-004-20107