Learn More
Medchemexpress LLC [D-Arg1,D-Phe5,D-Trp7,9,Leu11]-Substance P | 96736-12-8 | MFCD00076802 | 98.5% | 1516.83 g/mol | C79H109N19O12 | 5 MG

Supplier: Medchemexpress LLC HY1035445MG
[D-Arg1,D-Phe5,D-Trp7,9,Leu11]-Substance P is a synthetic peptide derivative of Substance P containing D-amino acid substitutions. It acts as a biased agonist at selected neuropeptide and chemokine receptors, modulates G-protein signaling (activates G12), influences MAPK pathways, and promotes neutrophil chemotaxis and Ca2+ mobilization.
- Peptide sequence: {d-Arg}-Pro-Lys-Pro-{d-Phe}-Gln-{d-Trp}-Phe-{d-Trp}-Leu-Leu-NH2
- Molecular formula: C79H109N19O12
- Molecular weight: 1516.83 g/mol
- Purity: 98.48% (HPLC)
- Typical applications: receptor signaling studies, biased agonism research, and neutrophil chemotaxis assays
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.