Learn More
Medchemexpress LLC D-arginine | 157-06-2 | MFCD00063116 | 98.0% | 174.20 g/mol | C6H14N4O2 | 1 G

Supplier: Medchemexpress LLC HYW0167811G
D-Arginine (H-D-Arg-OH) is the D-enantiomer of the amino acid arginine provided for research applications. Supplied as a solid and as aqueous solution, it serves as an inactive stereoisomer reference, analytical reagent, or control in biochemical and neurological studies.
- D-enantiomer of arginine.
- Research-grade reagent for analytical and biochemical studies.
- Purity 98.0%.
- Molecular formula C6H14N4O2; molecular weight 174.20 g/mol.
- Available as solid and 10 mM aqueous solution.
- Used as stereochemistry control and in neurological or myeloma research.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.