Learn More
Medchemexpress LLC D-arginine | 157-06-2 | MFCD00063116 | 98.0% | 174.20 g/mol | C6H14N4O2 | 5 G

Supplier: Medchemexpress LLC HYW0167815G
D-Arginine is the dextrorotatory enantiomer of the amino acid arginine. Provided as a research-grade solid with typical purity of 98.0%, it is used as a chemical reference standard, for stereochemical and biochemical studies, and in analytical applications that require the D-form.
- D-enantiomer of arginine
- Research-grade purity of 98.0%
- Molecular weight 174.20 g/mol
- Chemical formula C6H14N4O2
- Supplied as a solid in multiple sizes for laboratory use
- Suitable for stereochemical studies and analytical standards
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.