Learn More
Selleck Chemical LLC D-Arginine-E0654-500MG
SDP
Supplier:  Selleck Chemical LLC E0654500MG
Encompass_Preferred
D-Arginine is an inhibitor of arginine decarboxylase(ADC) to regulate putrescine biosynthesis
Catalog No. 50-313-7731