Learn More
Medchemexpress LLC D-asparagine monohydrate | 5794-24-1 | MFCD00149558 | >98.0% | 150.13 g/mol | C4H10N2O4 | 100 G

Supplier: Medchemexpress LLC HYW017444100G
D-Asparagine monohydrate is the D-enantiomer of the amino acid asparagine supplied as the crystalline monohydrate. It is intended as a biochemical reagent for research use, suitable as a reference standard or reagent in biochemical assays and amino-acid studies. Not for human or clinical use.
- High purity (≥98.0%) for analytical and assay applications.
- Molecular weight 150.13 g/mol and formula C4H10N2O4.
- White solid form, stable when stored protected from light.
- Storage recommendations for solutions: -80°C (6 months) or -20°C (1 month).
- Available in multiple pack sizes to suit laboratory workflows.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.