Learn More
Medchemexpress LLC D-Asparagine (Standard) | 2058-58-4 | 99.9% | 132.12 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW010378R100MG
Encompass_Preferred
D-Asparagine (Standard) (H-D-Asn-OH (Standard)) is the analytical standard of D-Asparagine. This product is intended for research and analytical applications. D-Asparagine (H-D-Asn-OH) is a competitive inhibitor of L-Asparagine hydrolysis with a Ki value of 0.24 mM. It serves as a source of nitrogen for yeast strains and is a good substrate for the external yeast asparaginase but a poor substrate for the internal enzyme.
  • Provides an analytical standard for D-Asparagine, useful for research and analytical applications.
  • Acts as a competitive inhibitor of L-Asparagine hydrolysis.
  • Serves as a nitrogen source for yeast strains.
Catalog No. 50-003-67039