Learn More
Selleck Chemical LLC D-Aspartic acid S5129-25mg
SDP
Supplier:  Selleck Chemical LLC S512925MG
Encompass_Preferred
D-Aspartic acid (D-Asp Aminosuccinic acid asparagic acid asparaginic acid) is an endogenous amino acid occurring in several tissues and cells of both invertebrates and vertebrates It can regulate testosterone synthesis and may act on a stimulatory receptor (NMDA)
Catalog No. 50-310-8509