Learn More
Cayman Chemical D-AspartIc AcId 25g
SDP
Supplier:  Cayman Chemical 3349425 G
Encompass_Preferred
An amino acid and a precursor in NMDA biosynthesis; binds to the L-glutamate site of ionotropic NMDA receptors (Ki = 1.6 µM); increases cAMP production in rat synaptosomes at 1 mM; enhances electrically stimulated fEPSPs in rat hippocampal slices; increases growth hormone, luteinizing hormone, testosterone, and progesterone serum levels in rats
Catalog No. 50-294-9463