Learn More
Medchemexpress LLC D-aspartic acid beta tert-butyl ester | 64960-75-4 | MFCD00077101 | 99.9% | 189.21 g/mol | C8H15NO4 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW015233
Encompass_Preferred
H-D-Asp(OtBu)-OH is a tert-butyl-protected D-aspartic acid derivative used as a building block in peptide synthesis and related amino-acid chemistry. It is supplied as a white to off-white solid with a reported molecular weight of 189.21 g/mol and documented analytical data (COA, HPLC, MS) for research use.
  • Protected D-aspartic acid suitable for solid-phase peptide synthesis.
  • Tert-butyl protection on the side-chain carboxylate enables selective deprotection strategies.
  • High purity (99.9%) with supporting certificate of analysis and analytical reports.
  • Stable as a powder with recommended cold storage for extended shelf life.
  • Provided with datasheet, safety data sheet, and analytical documentation for traceability.
Catalog No. 50-003-03018