Learn More
Medchemexpress LLC HY-13735D 10mg Medchemexpress, d-Atabrine dihydrochloride CAS:56100-41-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13735D10MG
Encompass_Preferred
Medchemexpress, HY-13735D 10mg d-Atabrine dihydrochloride CAS:56100-41-5 d-Atabrine dihydrochloride is an active enantiomer of quinacrine which displays antiprion activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2684