Learn More
Medchemexpress LLC HY-N3021 10mg Medchemexpress, D-chiro-Inositol CAS:643-12-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN302110MG
Encompass_Preferred
Medchemexpress, HY-N3021 10mg D-chiro-Inositol CAS:643-12-9 D-chiro-Inositol is an epimer of myo-inositol found in certain mammalian glycosylphosphatidylinositol protein anchors and inositol phosphoglycans possessing insulin-like bioactivity. D-chiro-Inositol is used clinically for the treatment of polycystic ovary syndrome (PCOS) and diabetes mellitus, which can reduce hyperglycemia and ameliorate insulin resistance [1] [2] [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3936