Learn More
Medchemexpress LLC D-Cycloserine | 68-41-7 | 99.7% | C3H6N2O2 | 5 G
SDP
Supplier:  Medchemexpress LLC HYB00305G
Encompass_Preferred
D-Cycloserine is an antibiotic that targets sequential bacterial cell wall peptidoglycan biosynthesis enzymes and possesses blood-brain barrier permeability. It functions as a partial NMDA agonist, which can contribute to improved cognitive functions. This compound is also utilized in research concerning multidrug-resistant tuberculosis.
  • Targets sequential bacterial cell wall peptidoglycan biosynthesis enzymes
  • Possesses blood-brain barrier permeability
  • Functions as a partial NMDA agonist
  • Contributes to improved cognitive functions
  • Utilized in research concerning multidrug-resistant tuberculosis
Catalog No. 50-004-32025