Learn More
Medchemexpress LLC D-cysteine | 921-01-7 | MFCD00066461 | 98.0% | 121.16 g·mol⁻¹ | C3H7NO2S | 25 G

Supplier: Medchemexpress LLC 25GHYW018555
D-cysteine is the D-enantiomer of the amino acid cysteine supplied as a white to off-white solid for biochemical research. It is used in studies of stereochemistry-dependent biological activity, microbial modulation, and neural progenitor cell proliferation. The material is provided with documented purity and stability data for laboratory and analytical use.
- High purity (98.0%) suitable for research applications.
- White to off-white solid appearance for easy handling.
- Molecular formula C3H7NO2S; molecular weight 121.16 g·mol⁻¹.
- Powder storage: -20°C (3 years) or 4°C (2 years); soluble aliquots stable at -80°C.
- Datasheet, certificate of analysis, and safety data sheet available for quality and safety information.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.