Learn More
Medchemexpress LLC D-Cysteine hydrochloride | 32443-99-5 | 157.62 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW002074500G
Encompass_Preferred
D-Cysteine hydrochloride is a cysteine derivative that is commercially used as an ergogenic supplement. It influences the secretion of anabolic hormones, supplies fuel during exercise, improves mental performance during stress-related tasks, and helps prevent exercise-induced muscle damage. It is recognized as a beneficial ergogenic dietary substance.
  • Solid appearance
  • White to off-white color
  • Ships at room temperature in continental US (may vary elsewhere)
  • Store at 4°C, sealed, away from moisture; in solvent, store at -80°C for 6 months or -20°C for 1 month (sealed, away from moisture)
Catalog No. 50-004-48862