Learn More
Medchemexpress LLC D-Cystine | 349-46-2 | 240.30 | 25 G

Supplier: Medchemexpress LLC 25GHYW001941
D-Cystine is the D-enantiomer of L-Cystine. It inhibits L-aspartate-β-semialdehyde dehydrogenase (ASADH) from *Escherichia coli*.
- Inhibits L-aspartate-β-semialdehyde dehydrogenase (ASADH) from *Escherichia coli*
- D-enantiomer of L-Cystine
- Suitable for powder storage at -20°C for 3 years, 4°C for 2 years
- Suitable for storage in solvent at -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.