Learn More
Medchemexpress LLC D-Cystine | 349-46-2 | MFCD00002610 | 240.30 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW001941500MG
Encompass_Preferred
D-Cystine is the D-enantiomer of L-Cystine. It inhibits L-aspartate-β-semialdehyde dehydrogenase (ASADH) from Escherichia coli. This product is for research use only and not for sale to patients.
  • D-enantiomer of L-Cystine
  • Inhibits L-aspartate-β-semialdehyde dehydrogenase (ASADH)
  • Effective against Escherichia coli
  • Suitable for research applications
  • Classified as a natural product
  • Exhibits anti-infection properties
  • Functions as an antibiotic
  • Considered a drug isomer
Catalog No. 50-004-47587