Learn More
Genscript Corporation D-dimer 18D4 mAb Mouse
Supplier:  Genscript Corporation V0140210
Encompass
D-dimer known as a fibrin degradation product is presented in blood after a blood clot is degraded by fibrinolysis Its concentration in blood increases when deep venous thrombosis (DVT) pulmonary embolism (PE) or disseminated intravascular coagulation (DIC) happens It serves as a useful maker for the diagnosis of these diseases
Catalog No. 50-000-57799