Learn More
Cusabio Technology LLC Mouse D-Dimer,D2D ELISA Kit
Supplier:  Cusabio Technology LLC CSBE13584M
Encompass_Preferred
This Mouse D2D ELISA Kit was designed for the quantitative measurement of Mouse D2D protein in serum, plasma, cell culture supernates, tissue homogenates. It is a Sandwich ELISA kit, its detection range is 31.25 pg/mL-2000 pg/mL and the sensitivity is 7.81 pg/mL. One kit contains 96 wells.
Catalog No. NC2952952