Learn More
Cayman Chemical D-ErItadenIn 5mg
SDP
Supplier:  Cayman Chemical 217475 MG
Encompass_Preferred
An inhibitor of SAAH (IC50 = 7 nM); inhibits growth of C. parvum parasites (MIC50 = 3 μM) without exhibiting cytotoxicity in HCT-8 cells (CC50 = >1 mM); dietary administration increases liver microsomal phosphatidylethanolamine concentration and decreases liver microsomal Δ6 desaturase activity and plasma cholesterol levels in rats at 50 mg/kg
Catalog No. 50-298-7373