Learn More
Medchemexpress LLC D-erythro-sphingosine | 123-78-4 | MFCD00036751 | 98.0% | 299.49 | C18H37NO2 | 5 G

Supplier: Medchemexpress LLC HY1010475G
D-erythro-Sphingosine is a naturally occurring sphingoid base used in biochemical and cell biology research. It modulates kinase activity and is commonly used to probe signaling pathways in vitro.
- Natural sphingoid base used in biochemical and cell biology research.
- Modulates kinase activity and inhibits protein kinase C at micromolar concentrations.
- High purity (98.0%) suitable for research applications.
- Solid powder form; molecular weight 299.49; formula C18H37NO2.
- Stable as a powder at -20°C for up to 3 years; store in solvent at -80°C for long-term stability.
- Available in laboratory-scale quantities, including 5 g packaging.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.