Learn More
Apexbio Technology LLC D-erythro-Sphingosine (synthetic) 123-78-4 100mg
SDP
Supplier:  Apexbio Technology LLC B6385100
Encompass_Preferred
D-erythro-Sphingosine (synthetic) is an amino alcohol featuring a long-chain unsaturated hydrocarbon backbone Predominant in mammalian systems sphingosine serves as a structural component of sphingolipids alongside its analog sphinganine Functionally sphingosine modulates a variety of cellular processes by acting as a bioactive signaling molecule notably influencing pathways involved in cell growth differentiation and apoptosis In biomedical research D-erythro-Sphingosine is widely utilized to investigate sphingolipid metabolism signal transduction mechanisms and the role of sphingolipid derivatives in physiological and pathological states
Catalog No. 50-000-92221