Learn More
Medchemexpress LLC D-Erythrose 100mg | 583-50-6 | 120.10 g/mol | C4H8O4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY116956100MG
Encompass_Preferred
D-erythrose (50% in water) is the D-enantiomer of the four-carbon aldose erythrose supplied as a 50% aqueous solution for research and biochemical assay use. Formula C4H8O4; molecular weight 120.10 g/mol; CAS 583-50-6.
  • Four-carbon aldose with formula C4H8O4
  • Supplied as a 50% (w/v) aqueous solution for ease of handling
  • Intended for biochemical assays and metabolic pathway studies
  • Available in milligram-scale pack sizes suitable for analytical work
  • Provided with an accompanying safety data sheet for laboratory handling
Catalog No. 50-002-27666