Learn More
Medchemexpress LLC D-Erythrose 250mg | 583-50-6 | 120.10 g/mol | C4H8O4 | 250 MG

Supplier: Medchemexpress LLC HY116956250MG
D-Erythrose is a four-carbon aldose sugar supplied as a 50% (w/w) aqueous solution for use in biochemical assays and metabolic pathway studies. It functions as an intermediate in the pentose phosphate pathway and is provided as a liquid reagent for research and reagent preparation.
- Supplied as a 50% aqueous solution.
- High purity (reported 99.73%).
- Colorless to light yellow liquid with density ≈1.05 g/cm³.
- Used in biochemical assays and metabolic pathway studies.
- Recommended storage: protect from light; long-term storage at -20°C.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.