Learn More
Medchemexpress LLC D-Erythrose-4-13C 5mg | 90913-08-9 | 121.10 g/mol | C3(13C)H8O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY116956S35MG
Encompass_Preferred
D-Erythrose-4-13C is the 13C-labeled form of D-erythrose supplied as a solution in water for research use. It serves as a tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS.
  • 13C-labeled D-erythrose for tracer and quantitation
  • Supplied as a solution in water at a defined concentration (0.107 M for listed sizes)
  • Available in small, pre-measured sizes suitable for analytical workflows (5 mg, 10 mg, 25 mg)
  • Molecular formula C3(13C)H8O4; molecular weight 121.10 g/mol
  • Intended for research use only
Catalog No. 50-003-07740