Learn More
Medchemexpress LLC D-Fructose-13C | 108311-21-3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN7092S110MG
Encompass_Preferred
D-Fructose-13C is the 13C labeled D-Fructose. D-Fructose (D(-)-Fructose) is a naturally occurring monosaccharide found in many plants. It is for research use only and not sold to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-19891