Learn More
Cayman Chemical D-Fructose-13C6 100mg
SDP
Supplier:  Cayman Chemical 30071100 MG
Encompass_Preferred
An internal standard for the quantification of D-fructose by GC- or LC-MS
Catalog No. 50-299-0303