Learn More
Medchemexpress LLC D-Fructose-13C6 | 201595-65-5 | 25 MG

Supplier: Medchemexpress LLC 25MGHYN7092S
D-Fructose-13C6 is a 13C labeled D-Fructose, a naturally occurring monosaccharide that targets endogenous metabolites and metabolic enzyme/protease pathways. It is primarily used as a laboratory chemical for the manufacture of substances and can serve as a tracer or internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
- Molecular weight: 186.11
- Molecular formula: 13C6H12O6
- Appearance: White to off-white solid
- Purity (HPLC): 99.89%
- Isotopic enrichment: 98.48%
- Soluble in water: 250 mg/mL
- Used as a tracer or internal standard for quantitative analysis
- Not classified as a hazardous substance or mixture
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.