Learn More
Medchemexpress LLC D-fructose-6-phosphate disodium monohydrate | 99.88% | 322.11 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY113407CR50MG
Encompass_Preferred
D-Fructose-6-phosphate disodium monohydrate is an analytical standard and an intermediate of glycolysis. It is also a precursor for the synthesis of amino sugars like galactosamine and glucosamine. This compound is used as a reference standard in assays and commonly employed in qualitative, quantitative, and methodological research experiments using techniques like HPLC, GC, and MS.
  • It is the analytical standard of D-Fructose-6-phosphate disodium monohydrate.
  • It is used as a reference standard in assays.
  • It is commonly employed in qualitative, quantitative, and methodological research experiments using techniques like HPLC, GC, and MS.
  • It is an endogenous metabolite.
  • It is also found in human gut microbiota metabolites, human blood, cellular cytoplasm, and saliva.
Catalog No. 50-004-36904